missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA586894

Product Code. 16393755

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Purity is > 95% by SDS-PAGE.

FOXM1 (forkhead box protein M1) is a transcriptional factor regulating the expression of cell cycle genes essential for DNA replication and mitosis. FOXM1 plays a role in the control of cell proliferation and DNA break repairs participating in the DNA damage checkpoint response.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXM1
Polyclonal
Unconjugated
FOXM1
AA408308; AW554517; BB238854; cell growth-inhibiting gene 27 protein; FGCP; FKH L16; Fkh16; FKHL 16; FKHL16; folate hydrolase (prostate-specific membrane antigen) 1; Folate hydrolase 1; FOLH; FOLH1; Folylpoly-gamma-glutamate carboxypeptidase; forkhead box M1; forkhead box protein M1; forkhead homolog 16; Forkhead like 16; Forkhead, drosophila, homolog-like 16; forkhead-related protein FKHL16; FOXM 1; FOXM 1c; FOXM1; FOXM1B; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; hepatocyte nuclear factor 3 forkhead homolog 11; HFH 11; HFH11; HFH-11; HFH-11B; HNF 3; HNF3; HNF-3; HNF-3/fork-head homolog 11; INS 1; INS1; INS-1; INS-1 winged helix; Membrane glutamate carboxypeptidase; mGCP; M-phase phosphoprotein 2; MPHOSPH 2; MPHOSPH2; MPM 2; Mpm2; MPM-2 reactive phosphoprotein 2; MPP 2; MPP2; MPP-2; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; PIG 29; PIG29; prostate specific membrane antigen variant F; Prostate-specific membrane antigen; PSM; PSMA; Pteroylpoly-gamma-glutamate carboxypeptidase; TGT3; transcription factor Trident; TRIDENT; WIN; Winged-helix factor from INS-1 cells; winged-helix transcription factor Trident
Rabbit
Antigen Affinity Chromatography
RUO
14235, 2305, 58921
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O08696, P97691, Q08050
FOXM1
A synthetic peptide corresponding to residues in Human FOXM1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.