missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxM1 Mouse anti-Human, Clone: CL12131, Novus Biologicals™
Mouse Monoclonal Antibody
£302.00 - £480.00
Specifications
| Antigen | FoxM1 |
|---|---|
| Clone | CL12131 |
| Dilution | Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 2 to 10 μg/mL, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18327872
|
Bio-Techne
NBP3-15129-25UL |
25 μg |
£302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18045504
|
Novus Biologicals
NBP3-15129-100UL |
100 μg |
£480.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FoxM1 Monoclonal antibody specifically detects FoxM1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FoxM1 | |
| Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 2 to 10 μg/mL, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FKHL16trident, forkhead box M1, forkhead box protein M1, Forkhead, drosophila, homolog-like 16, forkhead-like 16, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH11FOXM1B, HFH-11M-phase phosphoprotein 2, HNF-3, HNF-3/fork-head homolog 11, INS-1, MPHOSPH2, MPP-2, MPP2MPM-2 reactive phosphoprotein 2, PIG29, TGT3, Transcription factor Trident, TRIDENT, WIN, Winged-helix factor from INS-1 cells | |
| This antibody was developed against a synthetic peptide. Actual immunogen sequence is proprietary information. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12131 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Mitotic Regulators | |
| PBS (pH 7.2), 40% glycerol | |
| 2305 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title