missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXL2NB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566360
This item is not returnable.
View return policy
Description
Immunogen sequence: PALVKKRMPDA CTLGRAGIGL PKMCLHMAVR HSKAQKTGPG ILQQRQKPPA PRASGGPALL GKRRGCSEA Highest antigen sequence identity to the following orthologs - mouse 30%, rat 29%.
FOXL2NB is a protein coding gene.
Specifications
| FOXL2NB | |
| Polyclonal | |
| Unconjugated | |
| FOXL2NB | |
| C3orf72; FLJ43329 protein; FOXL2 neighbor; FOXL2 neighbor protein; FOXL2NB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 401089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZUU3 | |
| FOXL2NB | |
| Recombinant Human FOXL2NB. Recombinant protein control fragment (Product #RP-105165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction