missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Rabbit anti-Human, Mouse, Rat, Clone: 4H5X3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16231-100UL
This item is not returnable.
View return policy
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| FOXL2 | |
| Monoclonal | |
| Unconjugated | |
| BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOXL2 (P58012). | |
| 100 μg | |
| Epigenetics | |
| 668 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4H5X3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction