missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXL2 Recombinant Rabbit Monoclonal Antibody (4H5X3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538083
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, K-562, Mouse testis, Rat testis, Rat ovary Immunogen sequence: MMASYPEPED AAGALLAPET GRTVKEPEGP PPSPGKGGGG GGGTAPEKPD PAQKPPYSYV ALIAMAIRES AEKRLTLSGI YQYIIAKFPF YEKNKKGWQN.
FOXL2 is a winged helix/forkhead transcription factor that is highly conserved in human, goat, mouse, and certain aquatic species. Given its cross-species conservation, it appears to play a critical role evolutionarily, likely in ovarian differentiation in mammals and in ovarian somatic cell differentiation and in further follicle development and/or maintenance. Immunohistochemical data shows that FOXL2 is a nuclear protein specifically expressed in eyelids and in fetal and adult ovarian follicular cells. It does not undergo any major posttranslational modifications. FOXL2 is also found to be associated with blepharo-phimosis/ptosis/epicanthus inverses syndrome (BPES). There are two forms of BPES. Type I BPES shows eyelid abnormal-ities that are associated with ovarian failure. In type II BPES, only eyelid defects are found. Studies have shown that FOXL2 is expressed in the mesenchyme of developing mouse eyelids and in adult ovarian follicles.
Specifications
| FOXL2 | |
| Recombinant Monoclonal | |
| 1.28 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88470, P58012 | |
| FOXL2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOXL2 (P58012). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4H5X3 | |
| Unconjugated | |
| FOXL2 | |
| AU045128; Blepharophimosis; BPES; BPES1; epicanthus inversus; forkhead box L2; forkhead box protein L2; forkhead transcription factor; forkhead transcription factor FOXL2; forkhead transcription factor L2; Foxl2; LUSTR2; MGC14393; Pfrk; P-Frk; PINTO; pituitary forkhead factor; POF3; putative transcription factor foxl2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26927, 367152, 668 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction