Learn More
FOXL2 Antibody (CL12762) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18544F
Product Code. 30305006
  • £582.00 / 0.10mL
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FOXL2
Monoclonal
FITC
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3
This antibody was developed using a synthetic peptide derived from P58012, with the exact immunogen sequence remaining proprietary.
0.1 mL
Epigenetics
668
Store at 4⪚C in the dark.
IgG3
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12762
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified