missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Antibody (CL12762) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18544IR
This item is not returnable.
View return policy
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| FOXL2 | |
| Monoclonal | |
| DyLight 755 | |
| BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
| This antibody was developed using a synthetic peptide derived from P58012, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Epigenetics | |
| 668 | |
| Store at 4⪚C in the dark. | |
| IgG3 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12762 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction