missing translation for 'onlineSavingsMsg'
Learn More

FOXL1 Antibody (2F6), Novus Biologicals™

Product Code. 18332508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332508 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332508 Supplier Novus Biologicals Supplier No. H00002300M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXL1 Monoclonal antibody specifically detects FOXL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2F6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005241
Gene Alias FKH6forkhead box protein L1, FKHL11forkhead-like 11, forkhead box L1, Forkhead-related protein FKHL11, Forkhead-related transcription factor 7, FREAC7FREAC-7
Host Species Mouse
Immunogen FOXL1 (NP_005241, 132 a.a. ~ 240 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MFENGNYRRRKRKPKPGPGAPEAKRPRAETHQRSAEAQPEAGSGAGGSGPAISRLQAAPAGPSPLLDGPSPPAPLHWPGTASPNEDAGDAAQGAAAVAVGQAARTGDGP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2300
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.