missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXJ2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552596
This item is not returnable.
View return policy
Description
Immunogen sequence: ELMESLRQAE QKNWTLDQHH IANLCDSLNH FLTQTGHVPP QGGTHRPPAP ARIADSCALT SGKQESAMSQ VNSYGHPQAP HLYPGPSPMY PIPTQDSAGY NRPAHHMVPR PSVPPPGANE EIPDDFDWDL I Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
FOXJ2 is a fork head factor that is expressed in many adult tissues. In the embryo, FOXJ2 expression showed a very early onset during the cleavage stages of preimplantation development. It is capable of activating transcription from promoters containing its sites.
Specifications
| FOXJ2 | |
| Polyclonal | |
| Unconjugated | |
| FOXJ2 | |
| FHX; Fork head homologous X; forkhead box J2; forkhead box protein J2; Foxj2; FOXJ2 forkhead factor; RGD1565101 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55810 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P0K8 | |
| FOXJ2 | |
| Recombinant protein corresponding to Human FOXJ2. Recombinant protein control fragment (Product #RP-89290). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction