missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXI1 Mouse anti-Human, Clone: PCRP-FOXL1-1F8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-13886-20UG
This item is not returnable.
View return policy
Description
FOXI1 Monoclonal antibody specifically detects FOXI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein Array
Specifications
| FOXI1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q12951 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2299 | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array | |
| PCRP-FOXL1-1F8 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| FKHL10HFH3, forkhead box I1, forkhead box protein I1, forkhead-like 10, forkhead-related activator 6, Forkhead-related protein FKHL10, Forkhead-related transcription factor 6, FREAC-6, FREAC6FKH10, Hepatocyte nuclear factor 3 forkhead homolog 3, HFH-3, HNF-3/fork-head homolog 3, HNF-3/fork-head homolog-3, MGC34197 | |
| Recombinant fragment (around aa104-258) of human FOXI1 protein (exact sequence is proprietary) (Uniprot: Q12951) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction