missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXI1 Monoclonal Antibody (OTI5D3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526378
This item is not returnable.
View return policy
Description
FOXI1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| FOXI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12951 | |
| FOXI1 | |
| Human recombinant protein fragment corresponding to amino acids 111-350 of FOXI1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI5D3 | |
| Unconjugated | |
| FOXI1 | |
| 5830401E05Rik; Fkh10; FKHL10; Forkhead; forkhead box I1; forkhead box protein I1; forkhead homolog 10; forkhead-like 10; forkhead-related activator 6; forkhead-related protein FKHL10; Forkhead-related transcription factor 6; Foxi1; FREAC6; FREAC-6; Hepatocyte nuclear factor 3 forkhead homolog 3; HFH3; HFH-3; HNF-3/forkhead homolog 3; HNF-3/fork-head homolog 3; HNF-3/fork-head homolog-3; MGC34197 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2299 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction