missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXI1 Antibody (PCRP-FOXL1-1F8) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14224JF669
This item is not returnable.
View return policy
Description
FOXI1 Monoclonal antibody specifically detects FOXI1 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| FOXI1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| FKHL10HFH3, forkhead box I1, forkhead box protein I1, forkhead-like 10, forkhead-related activator 6, Forkhead-related protein FKHL10, Forkhead-related transcription factor 6, FREAC-6, FREAC6FKH10, Hepatocyte nuclear factor 3 forkhead homolog 3, HFH-3, HNF-3/fork-head homolog 3, HNF-3/fork-head homolog-3, MGC34197 | |
| Recombinant fragment (around aa104-258) of human FOXI1 protein (exact sequence is proprietary) (Uniprot: Q12951) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-FOXL1-1F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2299 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction