missing translation for 'onlineSavingsMsg'
Learn More

FOXI1 Antibody (PCRP-FOXL1-1F8), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18379395 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379395 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379395 Supplier Novus Biologicals Supplier No. NBP314224AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXI1 Monoclonal antibody specifically detects FOXI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXI1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-FOXL1-1F8
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FKHL10HFH3, forkhead box I1, forkhead box protein I1, forkhead-like 10, forkhead-related activator 6, Forkhead-related protein FKHL10, Forkhead-related transcription factor 6, FREAC-6, FREAC6FKH10, Hepatocyte nuclear factor 3 forkhead homolog 3, HFH-3, HNF-3/fork-head homolog 3, HNF-3/fork-head homolog-3, MGC34197
Host Species Mouse
Immunogen Recombinant fragment (around aa104-258) of human FOXI1 protein (exact sequence is proprietary) (Uniprot: Q12951)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2299
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.