missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXI1 Antibody (OTI1D4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70747R
This item is not returnable.
View return policy
Description
FOXI1 Monoclonal specifically detects FOXI1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| FOXI1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FOXI1 | |
| Recombinant protein fragment corresponding to amino acids 111-350 of human FOXI1 (NP_036320) produced in E.coli. | |
| 0.1 mL | |
| 2299.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| FKHL10HFH3, forkhead box I1, forkhead box protein I1, forkhead-like 10, forkhead-related activator 6, Forkhead-related protein FKHL10, Forkhead-related transcription factor 6, FREAC-6, FREAC6FKH10, Hepatocyte nuclear factor 3 forkhead homolog 3, HFH-3, HNF-3/fork-head homolog 3, HNF-3/fork-head homolog-3, MGC34197 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction