missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572644
This item is not returnable.
View return policy
Description
FOXH1 Polyclonal Antibody for Western Blot
FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
Specifications
| FOXH1 | |
| Polyclonal | |
| Unconjugated | |
| FOXH1 | |
| contains winged-helix domain; etID62177.12; fast; Fast1; Fast-1; FAST1 transcription factor; FAST2; fast-2; fe26b06; Forkhead activin signal transducer 1; forkhead activin signal transducer 2; forkhead activin signal transducer FAST; forkhead activin signal transducer-1; forkhead box H1; forkhead box protein H1; Foxh1; hFAST-1; schmalspur; Schmalspur protein; SUR; TGF-beta/activin signal transducer; transcription factor; uncle freddy; unf; wu:fe26b06; wu:fi30c08; wu:fi32b01 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57930 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9I9E1 | |
| FOXH1 | |
| KLH conjugated synthetic peptide between 313-334 amino acids from the Central region of DANRE foxh1. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction