missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534474
This item is not returnable.
View return policy
Description
A suggested positive control is human liver tissue lysate. PA5-34474 can be used with blocking peptide PEP-1516.
FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
Specifications
| FOXH1 | |
| Polyclonal | |
| Unconjugated | |
| FOXH1 | |
| contains winged-helix domain; etID62177.12; fast; Fast1; Fast-1; FAST1 transcription factor; FAST2; fast-2; fe26b06; Forkhead activin signal transducer 1; forkhead activin signal transducer 2; forkhead activin signal transducer FAST; forkhead activin signal transducer-1; forkhead box H1; forkhead box protein H1; Foxh1; hFAST-1; schmalspur; Schmalspur protein; SUR; TGF-beta/activin signal transducer; transcription factor; uncle freddy; unf; wu:fe26b06; wu:fi30c08; wu:fi32b01 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14106, 300054, 8928 | |
| 4°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O75593, O88621 | |
| FOXH1 | |
| A 19 amino acid peptide near the amino terminus of human FOXH1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction