missing translation for 'onlineSavingsMsg'
Learn More

FOXG1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18326730 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326730 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326730 Supplier Novus Biologicals Supplier No. NBP2866360.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXG1 Polyclonal specifically detects FOXG1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias BF-1, BF1HBF-2, BF2, BF-2, Brain factor 1, Brain factor 2, FHKL3, FKHL1forkhead-like 1, FKHL2, FKHL3, FKHL4, forkhead box G1, forkhead box G1A, forkhead box G1B, forkhead box G1C, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, forkhead-like 3, forkhead-like 4, Forkhead-related protein FKHL1, Forkhead-related protein FKHL2, Forkhead-related protein FKHL3, FOXG1A, FOXG1Bforkhead-like 2, FOXG1C, HBF-1, hBF-2, HBF-3, HFK1HBF-G2, HFK2HBF2, HFK3KHL2, oncogene QIN, QINFKH2forkhead box protein G1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human FOXG1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2290
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.