missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXG1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA19043
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
FOXG1B belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of FOXG1B has not yet been determined; however, it may play a role in the development of the brain and telencephalonThis gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.
Specifications
| FOXG1 | |
| Polyclonal | |
| Unconjugated | |
| FOXG1 | |
| 2900064B05Rik; Bf1; BF-1; BF1A; BF2; BF-2; brain factor 1; Brain factor 2; FHKL3; FKH2; FKHL1; FKHL2; FKHL3; FKHL4; forkhead box G1; forkhead box G1B; forkhead box protein G1; Forkhead box protein G1A; Forkhead box protein G1B; Forkhead box protein G1C; forkhead-like 1; forkhead-like 2; forkhead-like 3; forkhead-like 4; forkhead-like transcription factor BF-1; forkhead-related protein FKHL1; Forkhead-related protein FKHL2; Forkhead-related protein FKHL3; FOXG1; FOXG1A; FOXG1B; FOXG1C; HBF-1; HBF2; HBF-2; HBF-3; HBF-G2; Hfh9; Hfhbf1; HFK1; HFK2; HFK3; HNF-3/forkhead homolog, brain factor 1; KHL2; oncogene QIN; QIN; RATBF1A; transcription factor BF-1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 2290 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55316 | |
| FOXG1 | |
| Synthetic peptide corresponding to the C terminus amino acids HQNQGSSSNPLIH (aa 477-489). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction