missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXG1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81942MFV610
This item is not returnable.
View return policy
Description
FOXG1 Polyclonal antibody specifically detects FOXG1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| FOXG1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| BF-1, BF1HBF-2, BF2, BF-2, Brain factor 1, Brain factor 2, FHKL3, FKHL1forkhead-like 1, FKHL2, FKHL3, FKHL4, forkhead box G1, forkhead box G1A, forkhead box G1B, forkhead box G1C, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, forkhead-like 3, forkhead-like 4, Forkhead-related protein FKHL1, Forkhead-related protein FKHL2, Forkhead-related protein FKHL3, FOXG1A, FOXG1Bforkhead-like 2, FOXG1C, HBF-1, hBF-2, HBF-3, HFK1HBF-G2, HFK2HBF2, HFK3KHL2, oncogene QIN, QINFKH2forkhead box protein G1 | |
| FOXG1 antibody was raised against an 18 amino acid peptide near the center of human FOXG1. The immunogen is located within amino acids 160 - 210 of FOXG1. Amino Acid Squence: KEGEKKNGKYEKPPFSYN | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 2290 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction