missing translation for 'onlineSavingsMsg'
Learn More

FOXG1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18402595 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402595 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402595 Supplier Novus Biologicals Supplier No. NBP281942FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXG1 Polyclonal antibody specifically detects FOXG1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXG1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias BF-1, BF1HBF-2, BF2, BF-2, Brain factor 1, Brain factor 2, FHKL3, FKHL1forkhead-like 1, FKHL2, FKHL3, FKHL4, forkhead box G1, forkhead box G1A, forkhead box G1B, forkhead box G1C, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, forkhead-like 3, forkhead-like 4, Forkhead-related protein FKHL1, Forkhead-related protein FKHL2, Forkhead-related protein FKHL3, FOXG1A, FOXG1Bforkhead-like 2, FOXG1C, HBF-1, hBF-2, HBF-3, HFK1HBF-G2, HFK2HBF2, HFK3KHL2, oncogene QIN, QINFKH2forkhead box protein G1
Host Species Rabbit
Immunogen FOXG1 antibody was raised against an 18 amino acid peptide near the center of human FOXG1. The immunogen is located within amino acids 160 - 210 of FOXG1. Amino Acid Squence: KEGEKKNGKYEKPPFSYN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2290
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.