missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115247
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FOXF1.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development. [provided by RefSeq, Jul 2008].
Specifications
| FOXF1 | |
| Polyclonal | |
| Unconjugated | |
| Foxf1 | |
| ACDMPV; AI450827; FKHL5; forkhead box F1; forkhead box F1a; forkhead box protein F1; forkhead box protein F1; LOW QUALITY PROTEIN: forkhead box protein F1; Forkhead, drosophila, homolog-like 5; forkhead-related activator 1; Forkhead-related protein FKHL5; forkhead-related transcription factor 1; Foxf1; Foxf1a; FREAC1; Freac-1; Hepatocyte nuclear factor 3 forkhead homolog 8; Hfh8; HFH-8; HNF-3/forkhead homolog 8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15227, 2294, 687536 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q12946, Q61080 | |
| Foxf1 | |
| A synthesized peptide derived from human FOXF1(Accession Q12946), corresponding to amino acid residues E310-G360. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction