missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112911
This item is not returnable.
View return policy
Description
FOXE3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Forkhead Box Protein E3 (FOXE3, forkhead-related protein FKHL12, forkhead-related transcription factor 8) is a forkhead/winged helix transcription factor, which is expressed in the developing lens from the start of lens placode induction and becomes restricted to the anterior proliferating cells when lens fiber differentiation begins.
Specifications
| FOXE3 | |
| Polyclonal | |
| Unconjugated | |
| FOXE3 | |
| dyl; dysgenetic lens; FKHL12; forkhead box E3; forkhead box protein E3; forkhead, drosophila, homolog-like 12; forkhead-related protein FKHL12; Forkhead-related transcription factor 8; Foxe3; FREAC8; FREAC-8; Hepatocyte nuclear factor 3 forkhead homolog 7; HFH7; HFH-7; HNF3; rct; Rinshoken cataract | |
| Rabbit | |
| Protein G | |
| RUO | |
| 171302, 2301, 30923 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.9 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q13461, Q63250, Q9QY14 | |
| FOXE3 | |
| Recombinant Human Forkhead box protein E3 protein (4-74AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction