missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXE3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540519
This item is not returnable.
View return policy
Description
Peptide sequence: PEPPCCAAPD AAAAAFPPCA AAASPPLYSQ VPDRLVLPAT RPGPGPLPAE Sequence homology: Human: 100%.
Forkhead Box Protein E3 (FOXE3, forkhead-related protein FKHL12, forkhead-related transcription factor 8) is a forkhead/winged helix transcription factor, which is expressed in the developing lens from the start of lens placode induction and becomes restricted to the anterior proliferating cells when lens fiber differentiation begins.
Specifications
| FOXE3 | |
| Polyclonal | |
| Unconjugated | |
| FOXE3 | |
| dyl; dysgenetic lens; FKHL12; forkhead box E3; forkhead box protein E3; forkhead, drosophila, homolog-like 12; forkhead-related protein FKHL12; Forkhead-related transcription factor 8; Foxe3; FREAC8; FREAC-8; Hepatocyte nuclear factor 3 forkhead homolog 7; HFH7; HFH-7; HNF3; rct; Rinshoken cataract | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2301 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13461 | |
| FOXE3 | |
| Synthetic peptide directed towards the C-terminal of human FOXE3 (aa 235-284). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto