missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXE3 Antibody [Alexa Fluor™ 700], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-46303AF700
This item is not returnable.
View return policy
Description
FOXE3 Polyclonal antibody specifically detects FOXE3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| FOXE3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Alexa Fluor 700 | |
| ASMD, FKHL12forkhead, drosophila, homolog-like 12, forkhead box E3, Forkhead-related protein FKHL12, Forkhead-related transcription factor 8, FREAC-8, FREAC8forkhead box protein E3 | |
| forkhead box E3. (Uniprot# Q13461) | |
| 0.1 mL | |
| Neuroscience, Sensory Systems, Vision | |
| 2301 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction