missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FOXE1 Polyclonal specifically detects FOXE1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | FOXE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | FKHL15HFKL5, forkhead box E1 (thyroid transcription factor 2), forkhead box E2, Forkhead box protein E2, forkhead, drosophila, homolog-like 15, Forkhead-related protein FKHL15, HFKH4Thyroid transcription factor 2, TITF2HNF-3/fork head-like protein 5, TTF2forkhead box protein E1, TTF-2FOXE2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 184-373 of human FOXE1 (NP_004464.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?