missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549759
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total FOXE1 protein.
This intronless gene belongs to the forkhead family of transcription factors, which is characterized by a distinct forkhead domain. This gene functions as a thyroid transcription factor which likely plays a crucial role in thyroid morphogenesis. Mutations in this gene are associated with congenital hypothyroidism and cleft palate with thyroid dysgenesis. The map localization of this gene suggests it may also be a candidate gene for squamous cell epithelioma and hereditary sensory neuropathy type I.
Specifications
| FOXE1 | |
| Polyclonal | |
| Unconjugated | |
| FOXE1 | |
| FKHL15; forkhead box E1; forkhead box E1 (thyroid transcription factor 2); forkhead box E2; forkhead box protein E1; Forkhead box protein E2; forkhead, drosophila, homolog-like 15; forkhead-related protein FKHL15; Foxe1; FOXE2; HFKH4; HFKL5; HNF-3/fork head-like protein 5; NMTC4; Thyroid transcription factor 2; Titf2; TTF2; TTF-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110805, 192274, 2304 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00358, Q8R2I0 | |
| FOXE1 | |
| Synthesized peptide derived from internal of human TTF2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction