missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXD4L1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5114463

Product Code. 17199545

  • £437.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive controls that may be used include: A20 Cell Lysate, MCF7 Cell Slide.

This gene is a member of the forkhead/winged-helix (FOX) family of transcription factors with highly conserved FOX DNA-binding domains. Members of the FOX family of transcription factors are regulators of embryogenesis and may play a role in human cancer. This gene lies in a region of chromosome 2 that surrounds the site where two ancestral chromosomes fused to form human chromosome 2. This region is duplicated elsewhere in the human genome, primarily in subtelomeric and pericentromeric locations, thus mutiple copies of this gene have been found. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXD4L1
Polyclonal
Unconjugated
FOXD4L1
bA395L14.1; forkhead box D4 like 1; forkhead box D4-like 1; forkhead box protein D4-like 1; FOXD4L1; FOXD4-like 1; FOXD5
Rabbit
Antigen affinity chromatography
RUO
200350
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q9NU39
FOXD4L1
FOXD4L1 antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human FOXD4L1. The immunogen is located within the last 50 amino acids of FOXD4L1.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.