missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD4L1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41158G
This item is not returnable.
View return policy
Description
FOXD4L1 Polyclonal antibody specifically detects FOXD4L1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| FOXD4L1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 200350 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 488 | |
| bA395L14.1, forkhead box D4 like 1, forkhead box D4-like 1, forkhead box protein D4-like 1, FOXD4-like 1, FOXD5 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human FOXD4L1. The immunogen is located within the last 50 amino acids of FOXD4L1. Amino Acid Squence: LRQRLRSHQGRGAGRAPV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction