missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD4L5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575765
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
5 FOXD4-like proteins (FOXD4L2, FOXD4L3, FOXD4L4, FOXD4L5 and FOXD4L6) are encoded by a strongly repeated region of the 9q12 chromosome region. They are very similar and it is therefore difficult to provide a clear and unambiguous protein sequence. Our sequences are in agreement with HGNC nomenclature.
Specifications
| FOXD4L5 | |
| Polyclonal | |
| Unconjugated | |
| FOXD4L5 | |
| bA15J10.2; forkhead box D4-like 5; forkhead box protein D4-like 5; FOXD4L5; FOXD4-like 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 653427 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VV16 | |
| FOXD4L5 | |
| Synthetic peptide corresponding to residues in Human FOXD4L5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction