missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD4 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84027-0.1ML
This item is not returnable.
View return policy
Description
FOXD4 Polyclonal specifically detects FOXD4 in Mouse samples. It is validated for Western Blot.
Specifications
| FOXD4 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 2298 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| FKHL9MGC105106, forkhead box D4, forkhead box protein D4, forkhead, Drosophila, homolog-like 9, forkhead-like 9, forkhead-related activator 5, Forkhead-related protein FKHL9, Forkhead-related transcription factor 5, FOXD4A, FREAC-5, FREAC5FOXD4a, Myeloid factor-alpha | |
| The immunogen is a synthetic peptide directed towards the N terminal region of mouse FOXD4 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction