missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106539
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FOXD3.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1.
Specifications
| FOXD3 | |
| Polyclonal | |
| Unconjugated | |
| FOXD3 | |
| AIS1; CWH3; forkhead box D3; forkhead box protein D3; FOXD3; Genesis; hepatocyte nuclear factor 3 forkhead homolog 2; HFH2; HFH-2; HNF3/FH transcription factor genesis; HNF-3/forkhead homolog 2; VAMAS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15221, 27022, 29203 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| ∼1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61060, Q63245, Q9UJU5 | |
| FOXD3 | |
| A synthesized peptide derived from human FOXD3(Accession Q9UJU5), corresponding to amino acid residues N213-A263. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction