missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD3 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148629
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1.
Specifications
| FOXD3 | |
| Polyclonal | |
| Unconjugated | |
| FOXD3 | |
| AIS1; CWH3; forkhead box D3; forkhead box protein D3; FOXD3; Genesis; hepatocyte nuclear factor 3 forkhead homolog 2; HFH2; HFH-2; HNF3/FH transcription factor genesis; HNF-3/forkhead homolog 2; VAMAS2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 27022 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UJU5 | |
| FOXD3 | |
| E. coli-derived recombinant human FoxD3, Met1-Val140. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction