missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxD3 Antibody (5G9), Novus Biologicals™
Mouse Monoclonal Antibody
£184.00
Specifications
| Antigen | FoxD3 |
|---|---|
| Clone | 5G9 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
FoxD3 Monoclonal specifically detects FoxD3 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FoxD3 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 27022 | |
| Purified recombinant fragment of human FOXD3 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 5G9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Embryonic Stem Cell Markers, Stem Cell Markers | |
| AIS1, forkhead box D3, forkhead box protein D3, HFH2, HNF3/FH transcription factor genesis | |
| FOXD3 | |
| IgG1 | |
| Unpurified | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title