missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106981
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 86%; Guinea Pig: 86%; Horse: 79%; Human: 79%; Mouse: 100%; Rat: 100%.
FOXD2 belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined.
Specifications
| FOXD2 | |
| Polyclonal | |
| Unconjugated | |
| FOXD2 | |
| AI426778; FKHL17; forkhead box D2; forkhead box protein D2; forkhead, drosophila, homolog-like 17; forkhead-like 17; forkhead-related activator 9; forkhead-related protein FKHL17; Forkhead-related transcription factor 9; Foxd2; FREAC9; FREAC-9; Mesoderm/mesenchyme forkhead 2; Mf2; MF-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| O35392 | |
| FOXD2 | |
| Synthetic peptide directed towards the C-terminal region of mouse Foxd2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction