missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142065
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
Specifications
| FOXD1 | |
| Polyclonal | |
| Unconjugated | |
| FOXD1 | |
| AI385632; BF-2; brain factor 2; FKHL8; forkhead box D1; forkhead box protein D1; FORKHEAD BOX PROTEIN D1 (FORKHEAD-RELATED PROTEIN FKHL8) (FORKHEAD-RELATED TRANSCRIPTION FACTOR 4) (FREAC-4); Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; forkhead-related protein FKHL8; Forkhead-related transcription factor 4; Foxd1; FREAC4; FREAC-4; Hfh10; Hfhbf2; HFH-BF-2; HNF-3/forkhead homolog, brain factor 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2297 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q16676 | |
| FOXD1 | |
| Peptide with sequence SSVENFTARISNC (aa 453-465). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction