missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535145
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
Specifications
| FOXD1 | |
| Polyclonal | |
| Unconjugated | |
| FOXD1 | |
| AI385632; BF-2; brain factor 2; FKHL8; forkhead box D1; forkhead box protein D1; FORKHEAD BOX PROTEIN D1 (FORKHEAD-RELATED PROTEIN FKHL8) (FORKHEAD-RELATED TRANSCRIPTION FACTOR 4) (FREAC-4); Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; forkhead-related protein FKHL8; Forkhead-related transcription factor 4; Foxd1; FREAC4; FREAC-4; Hfh10; Hfhbf2; HFH-BF-2; HNF-3/forkhead homolog, brain factor 2 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 15229, 2297 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q16676, Q61345 | |
| FOXD1 | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human FOXD1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction