missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FoxC2 Monoclonal antibody specifically detects FoxC2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | FoxC2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3E3 |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FKHL14LD, forkhead box C2 (MFH-1, mesenchyme forkhead 1), forkhead, Drosophila, homolog-like 14, Forkhead-related protein FKHL14, Mesenchyme fork head protein 1, mesenchyme forkhead 1, MFH-1, MFH-1 protein, MFH1forkhead box protein C2, Transcription factor FKH-14 |
| Host Species | Mouse |
| Immunogen | Recombinant protein fragment corresponding to amino acids 1-280 of human FoxC2 (NP_005242) produced in E.coli. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?