missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568774
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintenance of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.
Specifications
| FOXC1 | |
| Polyclonal | |
| Unconjugated | |
| FOXC1 | |
| ARA; ch; congenital hydrocephalus; Fkh1; fkh-1; Fkhl7; forkhead box C1; forkhead box C1 protein; forkhead box protein C1; forkhead, drosophila, homolog-like 7; forkhead/winged helix-like transcription factor 7; forkhead-related activator 3; forkhead-related protein FKHL7; Forkhead-related transcription factor 3; Foxc1; FREAC3; FREAC-3; frkhda; HGNC:3800; IGDA; IHG1; IRID1; Mesoderm/mesenchyme forkhead 1; Mf1; MF-1; Mf4; myeloid factor-delta; RIEG3; Transcription factor FKH-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17300 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q61572 | |
| FOXC1 | |
| synthetic peptide directed towards the n terminal region of mouse FOXC1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction