missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FoxC1 Antibody (SR2414), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27713
This item is not returnable.
View return policy
Description
FoxC1 Monoclonal antibody specifically detects FoxC1 in Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| FoxC1 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| SR2414 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:20 | |
| ARA, FKHL7RIEG3, forkhead box C1, forkhead box protein C1, forkhead, drosophila, homolog-like 7, forkhead/winged helix-like transcription factor 7, forkhead-related activator 3, Forkhead-related protein FKHL7, Forkhead-related transcription factor 3, FREAC-3, FREAC3IRID1, IGDA, IHG1, myeloid factor-delta | |
| Recombinant protein of human FoxC1 (Uniprot #: Q12948) | |
| 100 μL | |
| Cancer | |
| 2296 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction