missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FoxC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29488
This item is not returnable.
View return policy
Description
FoxC1 Polyclonal antibody specifically detects FoxC1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation
Specifications
| FoxC1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| ARA, FKHL7RIEG3, forkhead box C1, forkhead box protein C1, forkhead, drosophila, homolog-like 7, forkhead/winged helix-like transcription factor 7, forkhead-related activator 3, Forkhead-related protein FKHL7, Forkhead-related transcription factor 3, FREAC-3, FREAC3IRID1, IGDA, IHG1, myeloid factor-delta | |
| between 275 and 325 | |
| 100 μg | |
| Cancer | |
| 2296 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction