missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXB1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518170
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, human, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.
Specifications
| FOXB1 | |
| Polyclonal | |
| Unconjugated | |
| Foxb1 | |
| C43; FKH5; forkhead box B1; forkhead box B1a; forkhead box B1b; forkhead box protein B1; Foxb1; Foxb1a; Foxb1b; Hfh-e5.1; HFKH-5; mesoderm/mesenchyme forkhead 3; Mf3; Transcription factor FKH-5; Twh | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 27023, 64290 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q64732, Q99853 | |
| Foxb1 | |
| Synthetic peptide sequence (TSPASALHSVAVH) corresponding to the C-terminus amino acids of FOXB1 (aa 313-325). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction