missing translation for 'onlineSavingsMsg'
Learn More

FOXB1 Antibody (PCRP-FOXB1-1B7), HRP, Novus Biologicals™

Product Code. 18369723
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369723 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369723 Supplier Bio-Techne Supplier No. NBP314021H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXB1 Monoclonal antibody specifically detects FOXB1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXB1
Applications ELISA
Classification Monoclonal
Clone PCRP-FOXB1-1B7
Conjugate HRP
Formulation PBS
Gene Alias FKH5HFKH-5, forkhead box B1, forkhead box protein B1, Transcription factor FKH-5
Host Species Mouse
Immunogen Recombinant full-length human FOXB1protein (Uniprot: Q99853)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27023
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.