missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FOXB1 Monoclonal antibody specifically detects FOXB1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | FOXB1 |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-FOXB1-1B7 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | FKH5HFKH-5, forkhead box B1, forkhead box protein B1, Transcription factor FKH-5 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human FOXB1protein (Uniprot: Q99853) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?