missing translation for 'onlineSavingsMsg'
Learn More

FOXA3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Product Code. 18311746 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18311746 20 μL 20µL
18393044 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18311746 Supplier Novus Biologicals Supplier No. NBP30389620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXA3 Polyclonal specifically detects FOXA3 in Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FKHH3, Fork head-related protein FKH H3, forkhead box A3, Forkhead box protein A3, hepatocyte nuclear factor 3, gamma, hepatocyte nuclear factor 3-gamma, HNF-3G, HNF-3-gamma, HNF3GTCF-3G, MGC10179, TCF3G, Transcription factor 3G
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOXA3 (NP_004488.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 3171
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.