Learn More
FOXA2, eFluor 660, clone: 3C10, eBioscience™
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. This gene has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified for this gene.
Specifications
Specifications
| Antigen | FoxA2 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 3C10 |
| Concentration | 0.2mg/mL |
| Conjugate | eFluor 660 |
| Formulation | PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 |
| Gene | FOXA2 |
| Gene Accession No. | Q9Y261 |
| Gene Alias | HNF3B |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.