missing translation for 'onlineSavingsMsg'
Learn More

FOXA1 Mouse anti-Human, Rat, Clone: FOXA1/1518, Abnova™

Mouse Monoclonal Antibody

Brand:  Abnova MAB14558.100ug

Product Code. 16092866

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mouse monoclonal antibody raised against partial recombinant human FOXA1.

  • High specificity
  • High reactivity
Specifications

Specifications

FOXA1
Monoclonal
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
P55317
FOXA1
Recombinant protein corresponding to amino acids 467-614 of human FOXA1.
100 μg
Primary
Human, Rat
Liquid
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
FOXA1/1518
Mouse monoclonal antibody raised against partial recombinant human FOXA1.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
HNF3A/MGC33105/TCF3A
Mouse
Protein A/G purification
RUO
3169
Store at 4°C.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)