missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA1 Monoclonal Antibody (A2E9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA544989
This item is not returnable.
View return policy
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.Specifications
FOXA1 | |
Monoclonal | |
2 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P55317 | |
FOXA1 | |
Synthetic peptide within N-terminal human FOXA1. | |
100 μL | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
A2E9 | |
Unconjugated | |
FOXA1 | |
Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2 | |
Mouse | |
Protein G | |
RUO | |
3169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |