missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA1 / HNF3A Mouse, Clone: FOXA1/1512, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3169-MSM2-P0
This item is not returnable.
View return policy
Specifications
| FOXA1 / HNF3A | |
| Nuclear | |
| FOXA1/1512 | |
| Unconjugated | |
| NP_004487 | |
| Mouse | |
| 79kDa | |
| Protein A/G Purified | |
| RUO | |
| 3169 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| FOXA1 | |
| Recombinant human FOXA1 protein fragment (aa467-614) | |
| HepG-2 or MCF-7 cells. Liver, Pancreas, Bladder, Prostate, Colon and Lung and Breast. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction