missing translation for 'onlineSavingsMsg'
Learn More

FOXA1 / HNF3A Mouse, Clone: FOXA1/1241, enQuireBio™

Mouse Monoclonal Antibody

£181.83 - £423.29

Specifications

Antigen FOXA1 / HNF3A
Clone FOXA1/1241
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15950178
missing translation for 'viewDocuments'
EnquireBio
3169-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
15960178
missing translation for 'viewDocuments'
EnquireBio
3169-MSM1-P1
100 μg
£423.29
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

FOXA1 / HNF3A
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
3169
Recombinant human FOXA1 protein fragment (aa467-614)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
79kDa
FOXA1/1241
Immunohistochemistry
Unconjugated
RUO
NP_004487
FOXA1
IgG2b κ
Protein A/G Purified
Nuclear
HepG-2 or MCF-7 cells. Liver, Pancreas, Bladder, Prostate, Colon and Lung and Breast.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.