missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Four-Neck Round-Bottom Flasks

Product Code. 11826154
Change view
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
2000 mL
3000 mL
4000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11826154 1000 mL Each
11836154 2000 mL Each
11846074 3000 mL Each
11816154 4000 mL Each
11826084 10,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11826154 Supplier Lenz Laborglasintrumente™ Supplier No. 03345470
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reaction flasks with center neck and with 3 parallel side necks

  • Acc. to DIN 12392 and in addition to DIN standard
  • With 3 parallel side necks
  • With conical ground joints
  • With inscription label
  • Material: DURAN™

Specifications

Description Center neck - NS 45/40, Side necks - NS 29/32
Capacity (Metric) 1000 mL
Material Borosilicate Glass (3.3)
No. per Pack 1
Product Type Reaction flasks
Diameter (Metric) Outer 131 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.